missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Pivalate (stabilized with HQ) 99.0+%, TCI America™

Catalog No. P0668500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0668500ML 500 mL
P066825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0668500ML Supplier TCI America Supplier No. P0668500ML

Chemical Identifiers

CAS 3377-92-2
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00059211
InChI Key YCUBDDIKWLELPD-UHFFFAOYSA-N
Synonym Pivalic Acid Vinyl Ester, Trimethylacetic Acid Vinyl Ester, Vinyl Trimethylacetate
PubChem CID 256591
IUPAC Name ethenyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OC=C

Specifications

Melting Point -82°C
Color Colorless
Boiling Point 112°C
Quantity 500 mL
UN Number 3272
Formula Weight 128.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Pivalate (stabilized with HQ)
EINECSNumber (2)-0761
TSCA Yes