missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Vinyl Coated Clamp with Metal Chain

Catalog No. S15131
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S15131 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15131 Supplier Eisco™ Supplier No. PRCMP21

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Vinyl Coated Clamp with Metal Chain provides precise pressure when gripping glass objects such as beakers, Erlenmeyer flasks, test-tubes, boiling (Florence) flasks, and burettes to avoid potential breakage.

  • Screw adjustment allows for precise pressure when gripping glass objects to avoid potential breakage
  • Vinyl coated fingers provide a strong, slip-free hold and allow for use in high temperature applications
  • Round extension arm allows the clamp to be rotated 360 degrees, as well as varying the distance from the support apparatus
  • Solid Zinc die cast construction and a high grade, nontoxic, powder coating
  • Construction encourages rust free & long lasting use for any laboratory application
  • Ideal for handling beakers, Erlenmeyer flasks, test-tubes, boiling (Florence) flasks, burettes
  • An additional holder is required to secure the clamp to a frame, rod, or other apparatus (not included)
  • Clamp is attached to a stainless steel connecting rod measuring 5.5 in.long and 10 mm in diameter
  • Includes an attached, high quality 19 in. chain

Specifications

Type Joint Clamp
Material Zinc, Stainless Steel
Includes Vinyl-Coated Chain (19 in.)
For Use With (Equipment) Glassware
Color Blue