Learn More
Vileda Professional™ SanTech™ Duo Polyester Laminated Mop, Sterile
For disinfecting floors, walls and ceilings. Vileda Professional SanTech Duo Polyester Laminated Mop, Sterile are designed for use in cleanrooms up to class ISO 5/GMP A and B/FED209D Class 100.
Supplier:  Vileda Professional™ 147545
Catalog No. 19200716
Only null left
Description
Description
  • Made of polyester laminated cleanroom foam
  • Adhesive-free lamination techniques used to prevent additional contamination to products
  • Better liquid absorbency
  • Suitable for bucket and pre-prepared method
  • Validated to the 10-6 SAL per ISO 11137
  • Individually double bagged with Lot # and Indicator Dot on each package
  • Certificate of Sterility available upon request
Specifications
Specifications
Polyester
Product Suggestions
Product Suggestions