Learn More
Vileda Professional™ Single 6 Gallon Bucket with Casters
Compact bucket systems for clean environments designed for effective cleaning. Vileda Professional™ Single 6 Gallon Bucket with Casters is perfect for use with all Roll-O-Matic™ Mops.
Supplier:  Vileda Professional™ 149622
Catalog No. 19200741
Only null left
Description
Description
  • Solid and stable polypropylene plastic buckets and bails are 100% autoclavable
  • Unique electro-polished sieve (sold separately) with hole pattern, integrated bucket splash guard and handle support
  • One 6 gallon bucket
  • Swivel plastic autoclavable casters, fixed to bucket with stainless steel pin connection
Product Suggestions
Product Suggestions