missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Staurosporine, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 62996-74-1 |
| Physical Form | Neat |
| Linear Formula | C28H26N4O3 |
| MDL Number | MFCD00077402 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 466.53 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH