missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Semicarbazide hydrochloride, MilliporeSigma™ Supelco™

Numéro de catalogue. 111019485
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111019485 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111019485 Fournisseur Merck Emd Millipore Code fournisseur 33656100MGR
Il en reste null
Pharmaceutical (small molecule)

Spécifications

Product Line VETRANAL
CAS 563-41-7
Physical Form Neat
Linear Formula NH2 CONHNH2 · HCl
MDL Number MFCD00013009
Melting Point 175°C to 177°C
Synonym N-Aminourea hydrochloride; Hydrazine carboxamide hydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 111.53
Grade Analytical Standard
Afficher plus de résultats Afficher moins de résultats

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH