missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Posaconazole, MilliporeSigma™ Supelco™

Catalog No. 111013331
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013331 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013331 Supplier Merck Emd Millipore Supplier No. 3210325MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Posaconazole is an azole antifungal drug used mostly against zygomycetes.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 171228-49-2
Physical Form Neat
Linear Formula C37H42F2N8O4
MDL Number MFCD00941162
Synonym 4-[4-[4-[4-[[(3 R,5 R)-5-(2,4-Difluorophenyl)-5-(1,2,4-triazol-1-ylmethyl)oxolan-3-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-[(2 S,3 S)-2-hydroxypentan-3-yl]-1,2,4-triazol-3-one
Type Analytical Standard
Quantity 25 mg
Formula Weight 700.78
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH