missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Phenazopyridine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111013006
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013006 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013006 Supplier Merck Emd Millipore Supplier No. 34076100MG
Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 136-40-3
Physical Form Neat
Linear Formula C11H11N5 · HCl
MDL Number MFCD00035347
Synonym 2,6-Diamino-3-(phenylazo)pyridine hydrochloride; Urodine
Type Analytical Standard
Quantity 100 mg
Formula Weight 249.70
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH