missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Meropenem trihydrate, MilliporeSigma™ Supelco™

Catalog No. 111012724
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012724 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012724 Supplier Merck Emd Millipore Supplier No. 3246025MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Meropenem trihydrate (Meropenem) is a broad-spectrum carbapenem with antibacterial properties. It inhibits cell wall synthesis in both gram-negative and gram-positive bacteria.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 119478-56-7
Shipping Condition Wet ice
Physical Form Neat
Linear Formula C17H25N3O5 S · 3H2 O
MDL Number MFCD08600005
Synonym (1 R,5 S,6 S)-2-[(3 S,5 S)-5-(dimethylaminocarbonyl)pyrrolidin-3-ylthio]-6-[(R)-1-hydroxyethyl]-1-methylcarbapen-2-em-3-carboxylic acid trihydrate
Type Analytical Standard
Quantity 25 mg
Formula Weight 437.51
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH