missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Medroxyprogesterone 17-acetate, MilliporeSigma™ Supelco™

Catalog No. 111015332
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015332 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015332 Supplier Merck Emd Millipore Supplier No. 46412250MG
Only null left
Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 71-58-9
Physical Form Neat
Linear Formula C24H34O4
MDL Number MFCD00010483
Melting Point 206°C to 207°C
Synonym 17 alpha-Acetoxy-6 alpha-methylprogesterone; 17 alpha-Hydroxy-6 alpha-methyl-4-pregnene-3,20-dione 17-acetate; 6 alpha-Methyl-17 alpha-acetoxyprogesterone; 6 alpha-Methyl-17 alpha-hydroxyprogesterone acetate
Type Analytical Standard
Quantity 250 mg
Formula Weight 386.52
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH