missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Malachite Green oxalate salt, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Spécifications
Spécifications
| Product Line | VETRANAL |
| CAS | 2437-29-8 |
| Physical Form | Neat |
| Linear Formula | C23H25N2 · C2 HO4 · 0.5C2H2O4 |
| Synonym | N,N,N′,N′-Tetramethyl-4,4 ′-diaminotriphenylcarbenium oxalate; Basic Green 4 |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 463.50 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH