missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Ketoprofen-d3, MilliporeSigma™ Supelco™

Catalog No. 111018506
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018506 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018506 Supplier Merck Emd Millipore Supplier No. 3267310MG
Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 159490-55-8
Physical Form Neat
Linear Formula C16D3H11O3
MDL Number MFCD08063452
Synonym 2-(3-Benzoylphenyl)propionic acid-(methyl-d3)
Type Analytical Standard
Quantity 10 mg
Formula Weight 257.30
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH