missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Febantel, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 58306-30-2 |
| Physical Form | Neat |
| Linear Formula | C20H22N4O6 S |
| MDL Number | MFCD01738527 |
| Synonym | N-{2-[2,3-Bis-(methoxycarbonyl)-guanido]-5-(phenylthio)-phenyl}-2-methoxyacetamide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 446.48 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH