missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Ethopabate, MilliporeSigma™ Supelco™

Catalog No. 111014780
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014780 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014780 Supplier Merck Emd Millipore Supplier No. 33996100MGR
Only null left

Ethopabate is an antibiotic used as a coccidiostat in poultry feeds.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 59-06-3
Physical Form Neat
Linear Formula C12H15 NO4
MDL Number MFCD00075809
Synonym Methyl 4-acetamido-2-ethoxybenzoate; Ethopabate; Ethyl pabate
Type Analytical Standard
Quantity 100 mg
Formula Weight 237.25
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH