missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ α-Estradiol, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 57-91-0 |
| Physical Form | Neat |
| Linear Formula | C18H24O2 |
| MDL Number | MFCD00064144 |
| Synonym | 1,3,5(10)-Estratriene-3,17 alpha-diol; 3,17 alpha-Dihydroxy-1,3,5(10)-estratriene; Epiestradiol |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 272.38 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH