missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Erythromycin A dihydrate, MilliporeSigma™ Supelco™

Catalog No. 111014788
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014788 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014788 Supplier Merck Emd Millipore Supplier No. 46256250MG
Only null left
Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 59319-72-1
Physical Form Neat
Linear Formula C37H67 NO13 · 2H2 O
MDL Number MFCD16661001
Type Analytical Standard
Quantity 250 mg
Formula Weight 769.96
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH