missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Dipyron monohydrate, MilliporeSigma™ Supelco™

Catalog No. 111014782
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014782 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014782 Supplier Merck Emd Millipore Supplier No. 46232250MG

Metamizol is an active pharmaceutical ingredient with both analgesic and antipyretic effects, primarily used to relieve pain caused by neuritis and neuralgia.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 5907-38-0
Physical Form Neat
Linear Formula C13H16N3 NaO4 S · H2O
MDL Number MFCD00150293
Synonym Metamizol; Methapyrone
Type Analytical Standard
Quantity 250 mg
Formula Weight 351.35
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH