missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Demeclocycline hydrochloride hydrate, MilliporeSigma™ Supelco™

Catalog No. 111015059
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015059 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015059 Supplier Merck Emd Millipore Supplier No. 46161100MG

Chemical structure: tetracycline

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 64-73-3
Physical Form Neat
Linear Formula C21H21 ClN2O8 · HCl · xH2 O
MDL Number MFCD00149217
Synonym Chlorodemethyltetracycline hydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 501.31 (anhydrous basis)
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH