missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Ceftiofur, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 80370-57-6 |
| Physical Form | Neat |
| Linear Formula | C19H17N5O7S3 |
| MDL Number | MFCD00864953 |
| Synonym | 3-[(2-Furylcarbonyl)thio]methyl]-7-[2-(2-amino-4-thiazolyl)-2-(methoxyiminoacetamido)-3-cephem-4-carboxylic acid |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 523.56 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH