missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Albendazole sulfoxide, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 54029-12-8 |
| Physical Form | Neat |
| Linear Formula | C12H15N3O3 S |
| MDL Number | MFCD00797922 |
| Synonym | Ricobendazole; Albendazole oxide; Albendazole sulfoxide; Methyl [5-(propane-1-sulfinyl)-1 H-benzoimidazol-2-yl]-carbamate |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 281.33 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH