Learn More
Applied Biosystems™ VetMAX™ Réactifs PRRSV 3.0
Description
VetMAX PRRSV 3.0 Reagents contain master mix, Xeno RNA control, and a primers/probes mix that enable multiplex, one-step, reverse transcription and real-time PCR (RT-PCR) assays targeting North American (NA) and European (EU) porcine reproductive and respiratory syndrome virus (PRSSV) RNA and Xeno RNA. The Xeno RNA control (10,000 copies/μL) is added during extraction as an internal positive control (IPC). These reagents contain an updated primer/probe mix compared to the similar VetMAX NA and EU PRRSV 2.0 Reagents.
Features of the VetMAX PRRSV 3.0 Reagents include:
- Rapid and cost-effective method for amplification of target sequences
- Quality reagents manufactured under ISO 9001, providing confidence in supply and quality
- From a brand with over 20 years in the DNA/RNA arena, with extensive bioinformatics capabilities and experience
PRSSV causes significant losses in the swine industry due to reproductive disorders, respiratory conditions, and growth retardation. PRRSV can be identified in blood and semen. Real-time PCR is the most sensitive method for analyzing RNA amplification.
Spécifications
Spécifications
| Concentration | 2X (mix maître), 10X (mélange amorce/sonde) |
| Contenu et stockage | Conserver à -10° à -30 °C. |
| Maladie | Virus du syndrome reproducteur et respiratoire porcin |
| Quantité | 100 réactions |
| Shipping condition (Condition d'expédition) | Expédié à -30 °C à -10 °C |
| Nb de réactions | 100 réactions |
| Type d’échantillon | Sérum, liquides buccaux, tissu, sperme, liquide de traitement, sang total |
Usage exclusivement réservé à la recherche. Ne pas utiliser dans des procédures de diagnostic.