missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™ PRRSV 3.0 Controls

Catalog No. A53170
Encompass
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53170 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A53170 Supplier Applied Biosystems™ Supplier No. A53170

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

VetMAX™ PRRSV 3.0 Controls are for use in multiplex, real-time reverse transcription PCR (RT-PCR) assays targeting North American and European porcine reproductive and respiratory syndrome virus (PRRSV) and Xeno RNA.

VetMAX PRRSV 3.0 Controls are for use in multiplex, real-time reverse transcription PCR (RT-PCR) assays targeting North American and European porcine reproductive and respiratory syndrome virus (PRRSV) and Xeno RNA. The controls come pre-mixed in one tube as a ready-to-use solution.

The included Xeno RNA Control is an internal positive control added to the lysis solution during RNA isolation to distinguish PCR inhibition, reaction failure, or human error from true negatives.

Specifications

Concentration 2.5X
Content And Storage Store at -10° to -30°C.
Disease Porcine Reproductive and Respiratory Syndrome Virus
Quantity 10 Reactions
Shipping Condition Shipped at -30°C to -10°C
No. of Reactions 10 Reactions

For Research Use Only. Not for use in diagnostic procedures.