missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™-Plus qPCR Master Mix

Catalog No. 4415327
Encompass
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4415327 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4415327 Supplier Applied Biosystems™ Supplier No. 4415327

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with VetMAX™ Reagents and VetMAX™ Controls.

The VetMAX™-Plus qPCR Master Mix is designed for multiplex amplification of animal pathogen DNA targets using a rapid qPCR strategy. The kit includes Xeno™ DNA Control, which serves as an internal positive control for DNA purification and qPCR. DNA is amplified using AmpliTaq Gold™ DNA Polymerase on a real-time PCR system.

Specifications

Content And Storage Store at -20°C. Includes 100 reactions with the following components: qPCR Master Mix, Xeno™ DNA Control, and Nuclease-free water.
Detection Method Primer-probe
Form Liquid
GC-Rich PCR Performance Low
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 Fast System, 7500 System
Passive Reference Dye ROX (Pre-mixed)
Product Line VetMAX
Product Type PCR Reagents and Detection Kit for Animal Health
Quantity 100 Reactions
Shipping Condition Dry Ice
Sufficient For 100 Reactions
For Use With (Application) Gene Expression
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Sample Type DNA
Show More Show Less