Learn More
Applied Biosystems™ VetMAX™ Fast Multiplex Mastermix, with ROX
VetMAX Fast Multiplex Master Mix with ROX is designed to be a fast-cycling-capable, multiplex-assay-compatible one-step RT-PCR master mix for animal molecular diagnostics.
$338.00 - $3,065.00
Specifications
| Concentration | 2X |
|---|---|
| Detection Method | Primer-probe |
| Form | Liquid |
| PCR Method | 1-step RT-qPCR |
| Polymerase | AmpliTaq™ Fast DNA Polymerase |
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|
A57081
|
Applied Biosystems™
A57081 |
100 Reactions |
Each for $338.00
|
|
|||||
|
A57305
|
Applied Biosystems™
A57305 |
500 Reactions |
Each for $1,586.00
|
|
|||||
|
A57306
|
Applied Biosystems™
A57306 |
1000 Reactions |
Each for $3,065.00
|
|
|||||
Description
VetMAX Fast Multiplex Master Mix with ROX is designed to be a fast-cycling-capable, multiplex-assay-compatible one-step RT-PCR master mix for testing in animal-derived samples. It can be used to amplify pathogenic DNA and RNA present in a variety of animal sample matrices, including ones with challenging PCR inhibitors. The fast-cycling capability, combined with multiplexing compatibility, allows for rapid and accurate real-time PCR results in <50 minutes.
Features of VetMAX Fast Multiplex Master Mix with ROX include:
- Minimize time to results while maximizing accuracy and analytical sensitivity; amplify both DNA and RNA targets on the same plate using fast cycling
- Improve flexibility by running up to four targets (not including the passive reference dye, ROX) on the same plate; run all assays using the same fast cycling thermal profile for improved efficiency
- Increase analytical sensitivity of assays; formulated for increased analytical sensitivity while still being fast-cycling capable and multiplex-assay compatible
- Strengthen reaction tolerance to inhibitors commonly found in animal-sourced sample matrices such as blood, feces, oral fluids, milk, and smegma
Fast and flexible
VetMAX Fast Multiplex Master Mix with ROX is ideal for fast-cycling, multiplex-capable molecular testing workflows. Due to its one-step RT-PCR capability, it can amplify both DNA and RNA targets using a single, fast-cycling thermal protocol that runs in <50 minutes.
Multiplex compatible
VetMAX Fast Multiplex Master Mix with ROX has been designed for multiplexing up to four targets (not including the passive reference channel, ROX). This meets the demand for more data per reaction and does so with increased analytical sensitivity and accuracy.
Robust and reliable
VetMAX Fast Multiplex Master Mix with ROX has been tested with animal-sourced samples of varying complexities ranging from simple samples like serum to complex samples like feces and porcine oral fluids. Its tolerance to PCR inhibition makes this product ideal for users who require workflow efficiency even when dealing with multiple sample types and pathogen assays.
Specifications
| 2X | |
| Liquid | |
| AmpliTaq™ Fast DNA Polymerase | |
| ROX | |
| 5 min. | |
| Mastermix | |
| DNA/RNA |
| Primer-probe | |
| 1-step RT-qPCR | |
| Fast | |
| Animal Health | |
| TaqMan™ | |
| Built-in Hot Start |
For Research Use Only. Not for use in diagnostic procedures.