missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™ Fast Multiplex Mastermix, with ROX

Catalog No. A57081
Change view
Click to view available options
No. of Reactions:
100 Reactions
1000 Reactions
500 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A57081 100 Reactions
A57305 500 Reactions
A57306 1000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A57081 Supplier Applied Biosystems™ Supplier No. A57081

VetMAX Fast Multiplex Master Mix with ROX is designed to be a fast-cycling-capable, multiplex-assay-compatible one-step RT-PCR master mix for animal molecular diagnostics.

VetMAX Fast Multiplex Master Mix with ROX is designed to be a fast-cycling-capable, multiplex-assay-compatible one-step RT-PCR master mix for testing in animal-derived samples. It can be used to amplify pathogenic DNA and RNA present in a variety of animal sample matrices, including ones with challenging PCR inhibitors. The fast-cycling capability, combined with multiplexing compatibility, allows for rapid and accurate real-time PCR results in <50 minutes.

Features of VetMAX Fast Multiplex Master Mix with ROX include:

  • Minimize time to results while maximizing accuracy and analytical sensitivity; amplify both DNA and RNA targets on the same plate using fast cycling
  • Improve flexibility by running up to four targets (not including the passive reference dye, ROX) on the same plate; run all assays using the same fast cycling thermal profile for improved efficiency
  • Increase analytical sensitivity of assays; formulated for increased analytical sensitivity while still being fast-cycling capable and multiplex-assay compatible
  • Strengthen reaction tolerance to inhibitors commonly found in animal-sourced sample matrices such as blood, feces, oral fluids, milk, and smegma

Fast and flexible

VetMAX Fast Multiplex Master Mix with ROX is ideal for fast-cycling, multiplex-capable molecular testing workflows. Due to its one-step RT-PCR capability, it can amplify both DNA and RNA targets using a single, fast-cycling thermal protocol that runs in <50 minutes.

Multiplex compatible

VetMAX Fast Multiplex Master Mix with ROX has been designed for multiplexing up to four targets (not including the passive reference channel, ROX). This meets the demand for more data per reaction and does so with increased analytical sensitivity and accuracy.

Robust and reliable

VetMAX Fast Multiplex Master Mix with ROX has been tested with animal-sourced samples of varying complexities ranging from simple samples like serum to complex samples like feces and porcine oral fluids. Its tolerance to PCR inhibition makes this product ideal for users who require workflow efficiency even when dealing with multiple sample types and pathogen assays.

Specifications

Concentration 2X
Detection Method Primer-probe
Form Liquid
Format Tube
PCR Method 1-step RT-qPCR
Polymerase AmpliTaq™ Fast DNA Polymerase
Reaction Speed Fast
Label or Dye ROX
Quantity 100 reactions
For Use With (Application) Animal Health
Activation Time 5 min.
Compatibility TaqMan™
Final Product Type Mastermix
Hot Start Built-in Hot Start
No. of Reactions 100 Reactions
Starting Material DNA/RNA
Volume 1 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.