Learn More
Kent Scientific VetFlo™ Vaporizer for Isoflurane with Funnel
Catalog No. 13005203
Encompass
Change view
Click to view available options
For Use With (Application):
Veterinary Use
Catalog No. For Use With (Application)
13005203 Veterinary Use
1 options
Catalog No. 13005203 Supplier Kent Scientific Supplier No. VETFLO1231
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For laboratory, veterinary anesthesia delivery.
  • Tech 3 high flow vaporizer
  • Lightweight
  • Temperature compensation
  • One-piece sump for added strength
  • Compact wicker system for increased drug capacity
Warranty
Three year (vaporizer)
Specifications
For Use With (Application) Veterinary Use
Type High Flow Vaporizer
For Use With (Equipment) Isoflurane (Tech 3) vaporizer
Includes Funnel
Material Aluminum
Dimensions (D x W x H) Stand: 25.4 x 30.48 x 33.02 cm (10 x 12 x 13 in.)