Learn More
STERIS Vesphene™ III se Disinfectant Cleaner
This Phenolic Disinfectant is the latest generation of high pH disinfectant specifically formulated for the routine disinfection of hard, non-porous surfaces.
Supplier: STERIS 647508CA
Description
- A liquid concentrate specifically formulated for the disinfection of hard, non-porous surfaces
- Phenolic active ingredients provide broad spectrum antimicrobial efficacy
- Use-dilution of rate of 0.8% v/v (1:128)
- Can be used alone or in rotation with the LpH™ III Disinfectants
- Ideal for use in a contamination control rotation, includes surfactants for surface cleaning while disinfecting
- Products include an extensive documentation package to meet validation objectives
- Supported by the STERIS Technical Services team for individualized application and validation assistance
- Each lot documented by Certificate of Analysis
Chemical Identifiers
| Phenolic Disinfectant | |
| Non-sterile | |
| 0.8% v/v (1:128) |
Specifications
| Phenolic Disinfectant | |
| 0.8% v/v (1:128) | |
| 4 x 1 gal./Cs. |
| Non-sterile | |
| Disinfecting Hard, Non-Porous Surfaces | |
| 1 gal. |