missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Vanadium (V) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$249.47 - $439.50
Chemical Identifiers
| CAS | 7803-55-6 |
|---|---|
| Molecular Formula | H4NO3V |
| Molecular Weight (g/mol) | 116.98 |
| MDL Number | MFCD00011430 |
| InChI Key | YBVKNHXQSRDWAA-UHFFFAOYSA-M |
| PubChem CID | 516859 |
| IUPAC Name | vanadiumoylol amine |
| SMILES | N.O[V](=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010058100
|
Ricca Chemical Company
RV010058100N |
100 mL |
Each for $249.47
|
|
|||||
|
RV010058500
|
Ricca Chemical Company
RV010058500N |
500 mL |
Each for $439.50
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 7803-55-6 | |
| 116.98 | |
| YBVKNHXQSRDWAA-UHFFFAOYSA-M | |
| vanadiumoylol amine |
| H4NO3V | |
| MFCD00011430 | |
| 516859 | |
| N.O[V](=O)=O |
Specifications
| Natural LDPE Bottle | |
| 95.81%,1.96%,0.23% | |
| 1.01 | |
| H4NO3V | |
| Approximately 0°C | |
| N.O[V](=O)=O | |
| 116.98 | |
| 516859 |