missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Vanadium (V) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010058100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010058100 100 mL
RV010058500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010058100 Supplier Ricca Chemical Company Supplier No. RV010058100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7803-55-6 , 7697-37-2 , 7732-18-5
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
PubChem CID 516859
IUPAC Name vanadiumoylol amine
SMILES N.O[V](=O)=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.81%, 1.96%, 0.23%
CAS Max % 99.73%, 2.04%, 0.23%
Specific Gravity 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity