missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Vanadium Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010564100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010564100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010564100 Supplier Ricca Chemical Company Supplier No. RV010564100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7803-55-6 , 7697-37-2 , 7732-18-5
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
PubChem CID 516859
IUPAC Name vanadiumoylol amine
SMILES N.O[V](=O)=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 96.02%, 1.96%, 0.02%
CAS Max % 99.94%, 2.04%, 0.02%
Specific Gravity 1
Physical Form Liquid
Color Colorless
pH <2
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade High Purity
Chemical Name or Material Vanadium ICP-MS Standard
Show More Show Less