missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Uranium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$644.00 - $1407.03

Chemical Identifiers

CAS 6159-44-0
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010325100
View Documents
Ricca Chemical Company
RV010325100N
100 mL
Each for $644.00
Only null left
 
RV010325250
View Documents
Ricca Chemical Company
RV010325250N
250 mL
Each for $977.03
Only null left
 
RV010325500
View Documents
Ricca Chemical Company
RV010325500N
500 mL
Each for $1,407.03
Only null left
 
Description

Description

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

6159-44-0

Specifications

Natural LDPE Bottle
95.86%,1.96%,0.18%
1
Approximately 0°C
100 mL
6159-44-0 , 7697-37-2 , 7732-18-5
99.78%,2.04%,0.18%
Liquid
Standard
High Purity
Videos
SDS
Documents

Documents