missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Uranium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| CAS Min % | 95.86%, 1.96%, 0.18% |
| CAS Max % | 99.78%, 2.04%, 0.18% |
| Specific Gravity | 1 |
| Physical Form | Liquid |
| Melting Point | Approximately 0°C |
| Type | Standard |
| Quantity | 100 mL |
| Grade | High Purity |