missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Tungsten Standard for ICP 1000 ppm in 4% NH3, Ricca Chemical
Description
- 1000 ppm in 4% NH3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 93.91%, 3.92%, 0.17% |
| CAS Max % | 97.75%, 4.08%, 0.17% |
| Specific Gravity | 1 |
| Physical Form | Liquid |
| Melting Point | Approximately 0°C |
| Type | Standard |
| Quantity | 500 mL |
| Grade | High Purity |