missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Titanium (Ti) Standard for AAS 1000 ppm in 5% HNO3/0.5% HF, Ricca Chemical

Catalog No. RV010056100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010056100 100 mL
RV010056500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010056100 Supplier Ricca Chemical Company Supplier No. RV010056100N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 5% HNO3/0.5% HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS Min % 92.21%, 4.90%, 0.49%, 0.40%
CAS Max % 95.97%, 5.10%, 0.51%, 0.42%
Specific Gravity 1.012
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity