missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Titanium Standard for ICP 10 g/L in 5% HCl/HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010423100N
Description
- 10 g/L in 5% HCl/HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Titanium ICP Standard | |
| 16962-40-6 , 7647-01-0 , 7664-39-3 | |
| 93.11%,5.10%,0.51% | |
| Liquid | |
| Colorless | |
| Approximately 0°C | |
| Miscible with water | |
| Standard | |
| High Purity |
| Natural LDPE Bottle | |
| 89.45%,4.90%,0.49% | |
| 1 | |
| UN3264 | |
| <2 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| 100 mL |