missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tin (Sn) Standard for AAS 1000 ppm in 20% HCl, Ricca Chemical

Catalog No. RV010083100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010083100 100 mL
RV010083500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010083100 Supplier Ricca Chemical Company Supplier No. RV010083100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 20% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7646-78-8 , 7647-01-0 , 7646-78-8
Molecular Formula Cl4Sn
Molecular Weight (g/mol) 260.51
InChI Key HPGGPRDJHPYFRM-UHFFFAOYSA-J
PubChem CID 24287
IUPAC Name tetrachlorostannane
SMILES Cl[Sn](Cl)(Cl)Cl

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.82%, 1.96%, 0.22%
CAS Max % 99.74%, 2.04%, 0.22%
Specific Gravity 1.112
Physical Form Liquid
UN Number UN1789
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity