missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Thulium Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010057500N

 View more versions of this product

Catalog No. RV010057500


Only null left
Explore available promotions

Description

Description

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
95.93%,1.96%,0.11%
1.01
O3Tm2
Approximately 0°C
[O--].[O--].[O--].[Tm+3].[Tm+3]
385.87
High Purity
12036-44-1,7697-37-2,12036-44-1
99.85%,2.04%,0.11%
Liquid
MFCD00011285
ZIKATJAYWZUJPY-UHFFFAOYSA-N
dithulium(3+) trioxidandiide
500 mL

Chemical Identifiers

12036-44-1
385.87
ZIKATJAYWZUJPY-UHFFFAOYSA-N
[O--].[O--].[O--].[Tm+3].[Tm+3]
O3Tm2
MFCD00011285
dithulium(3+) trioxidandiide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions