missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Thulium Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$569.59 - $1181.00
Chemical Identifiers
| CAS | 12036-44-1 |
|---|---|
| Molecular Formula | O3Tm2 |
| Molecular Weight (g/mol) | 385.87 |
| MDL Number | MFCD00011285 |
| InChI Key | ZIKATJAYWZUJPY-UHFFFAOYSA-N |
| IUPAC Name | dithulium(3+) trioxidandiide |
| SMILES | [O--].[O--].[O--].[Tm+3].[Tm+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010057100
|
Ricca Chemical Company
RV010057100N |
100 mL |
Each for $569.59
|
|
|||||
|
RV010057500
|
Ricca Chemical Company
RV010057500N |
500 mL |
Each for $1,181.00
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 12036-44-1 | |
| 385.87 | |
| ZIKATJAYWZUJPY-UHFFFAOYSA-N | |
| [O--].[O--].[O--].[Tm+3].[Tm+3] |
| O3Tm2 | |
| MFCD00011285 | |
| dithulium(3+) trioxidandiide |
Specifications
| Natural LDPE Bottle | |
| 95.93%,1.96%,0.11% | |
| 1.01 | |
| O3Tm2 | |
| Approximately 0°C | |
| [O--].[O--].[O--].[Tm+3].[Tm+3] | |
| 385.87 | |
| High Purity |
| 12036-44-1,7697-37-2,12036-44-1 | |
| 99.85%,2.04%,0.11% | |
| Liquid | |
| MFCD00011285 | |
| ZIKATJAYWZUJPY-UHFFFAOYSA-N | |
| dithulium(3+) trioxidandiide | |
| 100 mL |