CAS RN 12036-44-1
Thulium(III) oxide, REacton™, 99.997% (REO)
CAS: 12036-44-1 Molecular Formula: O3Tm2 Molecular Weight (g/mol): 385.87 MDL Number: MFCD00011285 InChI Key: ZIKATJAYWZUJPY-UHFFFAOYSA-N IUPAC Name: dithulium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Tm+3].[Tm+3]
Thulium ICP Standard, 1000 ppm Tm in 3% HNO3, Ricca Chemical
CAS: 12036-44-1
VeriSpec™ Thulium Standard for ICP/MS 10 ppm in 2% HNO3, Ricca Chemical
CAS: 12036-44-1 Molecular Formula: O3Tm2 Molecular Weight (g/mol): 385.87 MDL Number: MFCD00011285 InChI Key: ZIKATJAYWZUJPY-UHFFFAOYSA-N IUPAC Name: dithulium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Tm+3].[Tm+3]
Thulium AA Standard, 1000 ppm Tm in 3% HNO3, Ricca Chemical
CAS: 12036-44-1
VeriSpec™ Thulium Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical
CAS: 12036-44-1 Molecular Formula: O3Tm2 Molecular Weight (g/mol): 385.87 MDL Number: MFCD00011285 InChI Key: ZIKATJAYWZUJPY-UHFFFAOYSA-N IUPAC Name: dithulium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Tm+3].[Tm+3]
VeriSpec™ Thulium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 12036-44-1 Molecular Formula: O3Tm2 Molecular Weight (g/mol): 385.87 MDL Number: MFCD00011285 InChI Key: ZIKATJAYWZUJPY-UHFFFAOYSA-N IUPAC Name: dithulium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Tm+3].[Tm+3]
VeriSpec™ Thulium Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 12036-44-1 Molecular Formula: O3Tm2 Molecular Weight (g/mol): 385.87 MDL Number: MFCD00011285 InChI Key: ZIKATJAYWZUJPY-UHFFFAOYSA-N IUPAC Name: dithulium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Tm+3].[Tm+3]