missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Thiosulphate - S2O32- Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical

Catalog No. p-7074171
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010947250 250 mL
RV010947500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010947250 Supplier Ricca Chemical Company Supplier No. RV010947250C
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Facility

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7772-98-7 , 7732-18-5
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
PubChem CID 24477
IUPAC Name disodium sulfanidesulfonate
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Packaging Amber Glass Bottle
CAS Min % 97.86%, 0.14%
CAS Max % 100%, 0.14%
Specific Gravity 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 250 mL
Grade High Purity