missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Tellurium (Te) Standard for AAS 1000 ppm in 20% HCl, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$281.13 - $610.33
Chemical Identifiers
| CAS | 10026-07-0 |
|---|---|
| Molecular Formula | Cl4Te |
| Molecular Weight (g/mol) | 269.4 |
| InChI Key | SWLJJEFSPJCUBD-UHFFFAOYSA-N |
| PubChem CID | 61443 |
| SMILES | Cl[Te](Cl)(Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010085100
|
Ricca Chemical Company
RV010085100N |
100 mL |
Each for $281.13
|
|
|||||
|
RV010085500
|
Ricca Chemical Company
RV010085500N |
500 mL |
Each for $610.33
|
|
|||||
Description
- 1000 ppm in 20% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural LDPE Bottle | |
| 78.19%,19.60%,0.21% | |
| 1.02 | |
| Cl4Te | |
| Approximately 0°C | |
| Cl[Te](Cl)(Cl)Cl | |
| 100 mL | |
| High Purity |
| 10026-07-0 , 7647-01-0 , 7732-18-5 | |
| 81.39%,20.40%,0.21% | |
| Liquid | |
| UN1789 | |
| SWLJJEFSPJCUBD-UHFFFAOYSA-N | |
| 269.4 | |
| 61443 |