missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Tellurium Standard for ICP 1000 ppm in 20% HCl, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010320500N
Description
- 1000 ppm in 20% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 78.19%,19.60%,0.21% | |
| 1 | |
| Cl4Te | |
| Approximately 0°C | |
| SWLJJEFSPJCUBD-UHFFFAOYSA-N | |
| 269.4 | |
| 61443 |
| 10026-07-0 , 7647-01-0 , 7732-18-5 | |
| 81.39%,20.40%,0.21% | |
| Liquid | |
| UN1789 | |
| Standard | |
| Cl[Te](Cl)(Cl)Cl | |
| 500 mL | |
| High Purity |
Chemical Identifiers
| 10026-07-0 | |
| 269.4 | |
| 61443 |
| Cl4Te | |
| SWLJJEFSPJCUBD-UHFFFAOYSA-N | |
| Cl[Te](Cl)(Cl)Cl |