missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tellurium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010356100
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010356100 100 mL
RV010356250 250 mL
RV010356500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010356100 Supplier Ricca Chemical Company Supplier No. RV010356100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7803-68-1 , 7697-37-2 , 7732-18-5
Molecular Formula H6O6Te
Molecular Weight (g/mol) 229.642
InChI Key FXADMRZICBQPQY-UHFFFAOYSA-N
PubChem CID 62686
ChEBI CHEBI:30461
SMILES O[Te](O)(O)(O)(O)O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.89%, 1.96%, 0.15%
CAS Max % 99.81%, 2.04%, 0.15%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity