missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Strontium - Sr2+ Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical

Catalog No. RV010941100
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010941100 100 mL
RV010941250 250 mL
RV010941500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010941100 Supplier Ricca Chemical Company Supplier No. RV010941100N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Facility

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 1633-05-2 , 7732-18-5
Molecular Formula CO3Sr
Molecular Weight (g/mol) 147.63
MDL Number MFCD00011250
InChI Key LEDMRZGFZIAGGB-UHFFFAOYSA-L
PubChem CID 15407
IUPAC Name strontium(2+) carbonate
SMILES [Sr++].[O-]C([O-])=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 97.83%, 0.17%
CAS Max % 100%, 0.17%
Specific Gravity 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity