missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Strontium (Sr) Standard for AAS 2% Sr in 1% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$1418.57 - $3491.27
Chemical Identifiers
| CAS | 10042-76-9 |
|---|---|
| Molecular Formula | N2O6Sr |
| Molecular Weight (g/mol) | 211.63 |
| MDL Number | MFCD00011248 |
| InChI Key | DHEQXMRUPNDRPG-UHFFFAOYSA-N |
| PubChem CID | 24848 |
| IUPAC Name | strontium(2+) dinitrate |
| SMILES | [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010129100
|
Ricca Chemical Company
RV010129100N |
100 mL |
Each for $1,418.57
|
|
|||||
|
RV010129500
|
Ricca Chemical Company
RV010129500N |
500 mL |
Each for $3,491.27
|
|
|||||
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10042-76-9 | |
| 211.63 | |
| DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
| strontium(2+) dinitrate |
| N2O6Sr | |
| MFCD00011248 | |
| 24848 | |
| [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
| Natural LDPE Bottle | |
| 92.29%,4.73%,0.98% | |
| 1.01 | |
| N2O6Sr | |
| Approximately 0°C | |
| [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 211.63 | |
| 24848 |
| 10042-76-9,10042-76-9,7732-18-5 | |
| 96.05%,4.93%,1.02% | |
| Liquid | |
| MFCD00011248 | |
| DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
| strontium(2+) dinitrate | |
| 100 mL | |
| High Purity |