missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Strontium (Sr) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010084500
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010084500 500 mL
RV010084100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010084500 Supplier Ricca Chemical Company Supplier No. RV010084500N

missing translation for 'validMainframeMsg'

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10042-76-9 , 7697-37-2 , 7732-18-5
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
PubChem CID 24848
IUPAC Name strontium(2+) dinitrate
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 95.80%, 1.96%, 0.24%
CAS Max % 99.72%, 2.04%, 0.24%
Specific Gravity 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 500 mL
Grade High Purity