missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Sodium Standard for ICP 10 g/L in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010467250N
Description
- 10 g/L in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 92.41%,3.63%,1.96% | |
| 1 | |
| NNaO3 | |
| Approximately 0°C | |
| VWDWKYIASSYTQR-UHFFFAOYSA-N | |
| sodium nitrate | |
| 250 mL | |
| CHEBI:63005 |
| 7631-99-4,7631-99-4,7732-18-5 | |
| 96.19%,3.77%,2.04% | |
| Liquid | |
| MFCD00011119 | |
| Standard | |
| [Na+].[O-][N+]([O-])=O | |
| 84.99 | |
| 24268 | |
| High Purity |
Chemical Identifiers
| 7631-99-4 | |
| 84.99 | |
| VWDWKYIASSYTQR-UHFFFAOYSA-N | |
| CHEBI:63005 | |
| [Na+].[O-][N+]([O-])=O |
| NNaO3 | |
| MFCD00011119 | |
| 24268 | |
| sodium nitrate |