missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Sodium Standard for ICP 10 g/L in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$759.78 - $1815.13
Chemical Identifiers
| CAS | 7631-99-4 |
|---|---|
| Molecular Formula | NNaO3 |
| Molecular Weight (g/mol) | 84.99 |
| MDL Number | MFCD00011119 |
| InChI Key | VWDWKYIASSYTQR-UHFFFAOYSA-N |
| PubChem CID | 24268 |
| ChEBI | CHEBI:63005 |
| IUPAC Name | sodium nitrate |
| SMILES | [Na+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010467100
|
Ricca Chemical Company
RV010467100N |
100 mL |
Each for $759.78
|
|
|||||
|
RV010467250
|
Ricca Chemical Company
RV010467250N |
250 mL |
Each for $1,308.09
|
|
|||||
|
RV010467500
|
Ricca Chemical Company
RV010467500N |
500 mL |
Each for $1,815.13
|
|
|||||
Description
- 10 g/L in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 7631-99-4 | |
| 84.99 | |
| VWDWKYIASSYTQR-UHFFFAOYSA-N | |
| CHEBI:63005 | |
| [Na+].[O-][N+]([O-])=O |
| NNaO3 | |
| MFCD00011119 | |
| 24268 | |
| sodium nitrate |
Specifications
| Natural LDPE Bottle | |
| 92.41%,3.63%,1.96% | |
| 1 | |
| NNaO3 | |
| Approximately 0°C | |
| VWDWKYIASSYTQR-UHFFFAOYSA-N | |
| sodium nitrate | |
| 100 mL | |
| CHEBI:63005 |
| 7631-99-4,7631-99-4,7732-18-5 | |
| 96.19%,3.77%,2.04% | |
| Liquid | |
| MFCD00011119 | |
| Standard | |
| [Na+].[O-][N+]([O-])=O | |
| 84.99 | |
| 24268 | |
| High Purity |