missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Sodium - Na+ Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Facility

$316.16 - $600.84

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
PubChem CID 24268
ChEBI CHEBI:63005
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010940100
View Documents
Ricca Chemical Company
RV010940100N
100 mL
Each for $316.16
Only null left
 
RV010940250
View Documents
Ricca Chemical Company
RV010940250N
250 mL
Each for $421.72
Only null left
 
RV010940500
View Documents
Ricca Chemical Company
RV010940500N
500 mL
Each for $600.84
Only null left
 
Description

Description

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

7631-99-4
84.99
VWDWKYIASSYTQR-UHFFFAOYSA-N
CHEBI:63005
[Na+].[O-][N+]([O-])=O
NNaO3
MFCD00011119
24268
sodium nitrate

Specifications

Natural LDPE Bottle
97.64%,0.36%
1.01
NNaO3
Approximately 0°C
[Na+].[O-][N+]([O-])=O
84.99
24268
High Purity
7631-99-4 , 7732-18-5
100%,0.38%
Liquid
MFCD00011119
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium nitrate
100 mL
CHEBI:63005
Videos
SDS
Documents

Documents