missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Rhodium (Rh) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical
Description
- 1000 ppm in 2% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| CAS Min % | 92.90%, 4.90%, 0.20% |
| CAS Max % | 96.70%, 5.10%, 0.20% |
| Specific Gravity | 1.027 |
| Physical Form | Liquid |
| UN Number | UN3264 |
| Melting Point | Approximately 0°C |
| Quantity | 500 mL |
| Grade | High Purity |